Thermo Scientific™ SUN-SRi™ Phenolic Screw Thread Caps Available on GSA/VA Contract for Federal Government customers only.
Solid top with rubber liner
Manufacturer: Thermo Scientific™ 200 832
 View more versions of this product
Catalog No. 22-035114
  Check Availability
Add to Cart
    Specifications
    Material Phenolic
    Type Solid Top Screw Cap
    Material Closure Phenolic
    Material (Liner) PTFE/Rubber
    Thread Size 15-415
    Color Black
    GPI Thread 15-415
  • Description & Specifications
Related Products
Product Content Feedback