Thermo Scientific™ SUN-SRi™ Plate+™ Glass Coated Microplates

Chemical resistance similar to glass while retaining the advantages of polypropylene

Manufacturer: Thermo Scientific™ 400054

 View more versions of this product

Catalog No. 14-823-263

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    For Use With (Application) Pharmaceutical applications where polystyrene or polypropylene microplates are not applicable
    Format 96-well
    Material Polypropylene
    Surface Coating Silicone-di-oxide,Silicone-di-oxide
    Type 96-Well Microplate
    Volume (Metric) Well 300μL,300μL
    Volume (Metric) Working 50 to 290μL
    Well Design Round
    Bottom Plate Material Polypropylene
    Applications Pharmaceutical applications where polystyrene or polypropylene microplates are not applicable
    Well Shape Round

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit