Learn More
Thermo Scientific™ SUN-SRi™ Septa for 8-425 Screw Thread Caps Available on GSA/VA Contract for Federal Government customers only.
8mm PTFE/Silicone
Manufacturer:  Thermo Scientific™ 500 868
 View more versions of this product
Catalog No. 14-823-361
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
No
0.8cm
Yes
Standard Durometer Septa
8mm
0.314 in.
Red, White
PTFE/Silicone
Pre-Slit
0.045 in.