Thermo Scientific™ SUN-SRi™ SunFlo™ Dissolution Filters Available on GSA/VA Contract for Federal Government customers only.
Manufactured of Ultra High Molecular Weight (UHMW) foamed porous polyethylene
Manufacturer: Thermo Scientific™ 400 110
 View more versions of this product
Catalog No. 14-823-273
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) 1/8 in. cannula or probe
  • Description & Specifications
Related Products
Product Content Feedback