Learn More
Thermo Scientific™ SUN-SRi™ SunFlo™ Dissolution Filters
Manufactured of Ultra High Molecular Weight (UHMW) foamed porous polyethylene
Supplier: Thermo Scientific™ 400 107
Description
- Full flow design removes small particulates
- Fit securely on the end of a 1/8 in. cannula or sample probe
Specifications
70 μm | |
100 Pack | |
1/8 in. Cannula or Probe |