Learn More
Thermo Scientific™ SUN-SRi™ “To Contain” Volumetric Pipet
Sun SRI volumetric pipets allow for quick and precise transfer of solutions for confidence in results every time.
Manufacturer:  Thermo Scientific™ 205 044
 View more versions of this product
Catalog No. 14-823-239
Add to cart
Description
Description
Specifications
Specifications
Volumetric Pipet
±0.02 mL
ASTM E969
A
10 mL
To Contain
Borosilicate Glass