Learn More
Thermo Scientific™ SUN-SRi™ “To Contain” Volumetric Pipet
Sun SRI volumetric pipets allow for quick and precise transfer of solutions for confidence in results every time.
$582.50 - $637.50
Specifications
Calibration To Contain
Certifications/Compliance ASTM E969
Class A
Material Borosilicate Glass
Type Volumetric Pipet
View More Specs
Products 2
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
14-823-237
View Documents
Thermo Scientific™
205 024
5 mL
Case of 12 for $582.50
Add to cart
14-823-239
View Documents
Thermo Scientific™
205 044
10 mL
Case of 12 for $637.50
Add to cart
Description
Description
Specifications
Specifications
To Contain
A
Volumetric Pipet
±0.01 mL
ASTM E969
Borosilicate Glass
12/Cs.