Learn More
Thermo Scientific Thermo-Seal Heat Sealing Foil
Suitable for long-term, low-temperature storage, transportation, and high-temperature applications such as PCR
$136.00 - $1924.00
Catalog Number | Mfr. No. | Description | Format | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Format | Price | Quantity & Availability | ||||
AB-3559
|
Thermo Scientific
AB3559 |
ThermoSeal,foil laminate,Roll,78mmx370m |
Case for $1,924.00
|
|
AB-0559
|
Thermo Scientific
AB0559 |
Thermo-Seal; foil laminate, 85x135mm |
Case of 100 for $136.00
|
|
Description
- Can be used in waterbath thermal cyclers
- Foil/polypropylene laminate
- Provides very strong seal that can be peeled only with the Foil Stripper
- Can be pierced with a single pipet tip or pierce plate
- Excellent chemical resistance
- Seal Integrity Range: -80°C to +120°C