Thermo Scientific™ Thermolyne™ Benchtop Muffle Furnaces
Compact, lightweight benchtop furnaces
Manufacturer: Thermo Scientific™ F47910
 View more versions of this product
Catalog No. 10-554-3
  Check Availability
Add to Cart
    Specifications
    Capacity (Metric) 2L
    Temperature Range (Metric) 100 to 1200°C
  • Description & Specifications
Related Products
Product Content Feedback