Learn More
Thermo Scientific™ Thermolyne™ Heavy Duty Muffle Furnaces
Iideal for the most demanding applications, including ashing organics, heat treating tools and ties and heat treating glass.
Manufacturer:  Thermo Scientific™ FA1730
Catalog No. 10-557
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Requires : Electrical connection requires fixed wiring (no power cord)
Description
Description
  • Heavy-duty firebrick insulation is hard, abrasion-resistant and designed to support heavy loads
  • Heavy-duty steel case supports the most demanding requirements
  • Door is filled with refractory cement for long life
  • F1600 and F1700 models have heating elements embedded in special refractory cement for long service life and protection from harmful materials and mechanical shock
  • Heating elements on all four chamber sides ensure maximum heat transfer and chamber uniformity
  • Ceramic hearth is included to protect the bottom heating element
Choice of Controllers
  • Furnaces that use any of these three controls also use a mechanical Overtemperature Protection relay
  • B1: Digital single setpoint control with a single ramp to setpoint and dwell; single display shows actual temperature or setpoint
  • C1: Digital programmable control with 1 stored program of 8 segments
  • D1: Digital programmable control with 4 stored programs, 16 segments per program, and RS-232 communications interface