Learn More
Thermo Scientific™ Thermolyne™ Incubator Microtube Blocks Available on GSA/VA Contract for Federal Government customers only.
For modular block dri-baths
Manufacturer:  Thermo Scientific™ BK165X17AQ
Catalog No. 11-716-59Q
Add to cart
This item is not returnable. View return policy
Description
Description
Solid anodized aluminum resists harshest environments, provides maximum heat retention
  • 20 x 1.5mL capacity
  • Includes thermometer well
  • For all modular block dri-baths
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
2 in.
3 in.
Anodized Aluminum
5.2cm
7.8cm
Modular block dri-baths