Thermo Scientific™ Tips for Tecan™ and Axon™ Instruments PatchXpress Workstations Available on GSA/VA Contract for Federal Government customers only.

For use with Tecan™ Genesis™, Freedom Evo™, Cavro™, PatchXpress Workstations

Manufacturer: Thermo Scientific™ 902262XP

 View more versions of this product

Catalog No. 21-402-416

  Check Availability
Add to Cart

    Specifications

    Product Type Filtered Robotic Tip
    Material Polypropylene
    Quantity 96 Tips/Rack
    For Use With (Equipment) Tecan™ Genesis™, Freedom Evo™, Cavro™, patchXpress workstations
    Sterility Nonsterile
    Autoclavable Yes
    Tip Style Racked
    Packaging Rack
    Certifications/Compliance DNase, RNase, pyrogen free
    Volume (Metric) 50μL
    Color White

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit