Thermo Scientific™ TRACE™ TR-1 GC Columns Available on GSA/VA Contract for Federal Government customers only.
Manufacturer: Thermo Scientific™ 260A298P
Catalog No. 03-152-604
  Check Availability
Add to Cart
    Specifications
    Stationary Phase TR-1
    Diameter (Metric) Inner 0.53mm
    Length (Metric) 30m
  • Description & Specifications
Related Products
Product Content Feedback