Learn More
Thermo Scientific™ TruNarc™ Available on GSA/VA Contract for Federal Government customers only.
Narcotics officers, customs, border patrol and other law enforcement personnel can scan a sample for multiple narcotics in one single test
$25877.80 - $28025.00
Products 3
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
17-730-101 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
8000102101
One year warranty; Heroin test kit
Each for $25,877.80
Add to cart
17-730-103 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
8000102201
Two year warranty; Heroin test kit
Each for $27,997.11
Add to cart
17-730-102 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
8000101201
Two year warranty
Each for $28,025.00
Add to cart
Description
Description
  • Fast, accurate identification — Returns results in seconds. Based on lab-proven Raman spectroscopy
  • Easy to use — Intuitive menu-driven interface enables fast training and proficiency
  • Single test, multiple narcotics — Conduct a single analysis to determine if a narcotic is present, saving time and money
  • Non-contact sampling — Scan directly through plastic or glass to minimize contamination, reduce exposure and preserve evidence
  • Automated, tamper-proof records — Captures all scan results including time and date stamp and self check results to help expedite prosecution
  • TruNarc v1.3 software upgrade — Identifies small amounts of synthetic cannabinoid on leaf material - typically the final form found before consumption, as well as several priority substances including Phenethylamines, PCP and APAAN
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.