Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific™ TX-400 Rotor Adapters
For use with TX-400 Swinging Bucket Rotor
The adapters enable rotor to accept bottles up to 250mL and tubes to 100mL.
$378.00 - $594.00
  • Products 16
  • Description & Specifications
Products
Catalog Number Mfr. No. Description Price Quantity Availability  
75-003-788
Available on GSA/VA Contract for Federal Government customers only. 
75003788
175/200/225/250mL Bottle
Each for $395.00
Check Availability
Add to Cart
75-003-708
Available on GSA/VA Contract for Federal Government customers only. 
75003708
100mL Round-bottom, Open-tube
Each for $380.00
Check Availability
Add to Cart
75-003-707
Available on GSA/VA Contract for Federal Government customers only. 
75003707
50mL DIN Round-bottom
Each for $378.00
Check Availability
Add to Cart
75-003-799
Available on GSA/VA Contract for Federal Government customers only. 
75003799
50mL Round-bottom
Each for $379.00
Check Availability
Add to Cart
75-003-683
Available on GSA/VA Contract for Federal Government customers only. 
75003683
50mL Conical
Each for $390.00
Check Availability
Add to Cart
75-003-703
Available on GSA/VA Contract for Federal Government customers only. 
75003703
30/25mL DIN Round/Flat-bottom
Each for $386.00
Check Availability
Add to Cart
75-003-706
Available on GSA/VA Contract for Federal Government customers only. 
75003706
25mL Universal
Each for $378.00
Check Availability
Add to Cart
75-003-682
Available on GSA/VA Contract for Federal Government customers only. 
75003682
15mL Conical
Each for $395.00
Check Availability
Add to Cart
75-003-794
Available on GSA/VA Contract for Federal Government customers only. 
75003794
15mL Blood Collection (17 x 125mm)
Each for $378.00
Check Availability
Add to Cart
75-003-704
Available on GSA/VA Contract for Federal Government customers only. 
75003704
15mL Round-bottom Sarstedt
Each for $388.00
Check Availability
Add to Cart
75-003-798
Available on GSA/VA Contract for Federal Government customers only. 
75003798
14mL Conical Urine
Each for $388.00
Check Availability
Add to Cart
75-003-681
Available on GSA/VA Contract for Federal Government customers only. 
75003681
10mL Blood Collection or 15mL Corex/Kimble
Each for $390.00
Check Availability
Add to Cart
75-003-680
Available on GSA/VA Contract for Federal Government customers only. 
75003680
5/7mL Blood Collection
Each for $388.00
Check Availability
Add to Cart
75-003-793
Available on GSA/VA Contract for Federal Government customers only. 
75003793
3mL RIA or Round-bottom (without cap)
Each for $382.00
Check Availability
Add to Cart
75-003-700
Available on GSA/VA Contract for Federal Government customers only. 
75003700
1.5/2.0mL Conical Microtube
Each for $388.00
Check Availability
Add to Cart
75-003-825
Available on GSA/VA Contract for Federal Government customers only. 
75003825
4.5/6mL Greiner Blood Tubes
Pack of 4 for $594.00
Check Availability
Add to Cart
Descriptions & Specifications
Related Products