Thermo Scientific™ ViewPoint Enterprise Plug-Ins and Software Available on GSA/VA Contract for Federal Government customers only.
Plug-ins and Software to be used with the ViewPoint Appliance. The platform is the base for a complete customizable solution for remote monitoring and remote real-time command and control; capable of integrating radiation, environmental, and general purpose detectors.
Manufacturer: Thermo Scientific™ SYS/1/70870/903
 View more versions of this product
Catalog No. 19-177-226
  Check Availability
Add to Cart
Required Accessories : <p><b>System Specifications</b><br/>Operates with WindowsTM NT/2000/XP<br/></p><b>Client Software Requirements</b><ul><li>Microsoft WindowsTM NT 4 Service Pack 6 or</li><li>Microsoft WindowsTM 2000 Service Pack 4 or </li><li>Microsoft WindowsTM XP Service Pack 1 or higher</li></ul></p>
    Specifications
    Operating System Windows™ NT/2000/XP
    For Use With (Application) Real time personnel dosimetry and data management, area monitoring, integrated remote monitoring, instrument management, first responder environmental monitoring, site process monitoring
    Product Type View Point Enterprise Software
    Plug-Ins
    Each instrument, detector or sensor, which is capable of being remotely monitored by ViewPoint, has a unique Input Device Plug-In. This capability allows the instrument to communicate with ViewPoint Data Engine/System.
    Client Plug-Ins
    The ViewPoint User Interface and Data Engine are compatible with third-party Remote Monitoring Software systems. Compatability includes such systems as: MGPI-Teleview™, Industrial Innovative Solutions (IIS) – RADS™, and Bartlett – RMS™, and ESRI ArcView™ version 3.3 and 8. All purchased plug-ins are preloaded on the ViewPoint Appliance prior to delivery. Additional Input Device plug-ins, Client plug-ins, and User/Workstation License Bundles can be purchased and added to the original ViewPoint appliance via a patch file and updated security key
    • Real-time personnel dosimetry and data management
    • Area monitoring
    • Integrated remote monitoring
    • Instrument management
    • First Responder environmental monitoring
    • Site Process Monitoring
  • Description & Specifications
Related Products
Product Content Feedback