Learn More
Thermo Scientific™ ViewPoint Enterprise Plug-Ins and Software
Plug-ins and Software to be used with the ViewPoint Appliance. The platform is the base for a complete customizable solution for remote monitoring and remote real-time command and control; capable of integrating radiation, environmental, and general purpose detectors.
Supplier: Thermo Scientific™ SYS/1/70853/054
Requires :
System Specifications
Operates with Windows™ NT/2000/XP
- Microsoft Windows™ NT 4 Service Pack 6 or
- Microsoft Windows™ 2000 Service Pack 4 or
- Microsoft Windows™ XP Service Pack 1 or higher
Description
Plug-Ins
Each instrument, detector or sensor, which is capable of being remotely monitored by ViewPoint, has a unique Input Device Plug-In. This capability allows the instrument to communicate with ViewPoint Data Engine/System.
Client Plug-Ins
The ViewPoint User Interface and Data Engine are compatible with third-party Remote Monitoring Software systems. Compatability includes such systems as: MGPI-Teleview™, Industrial Innovative Solutions (IIS) – RADS™, and Bartlett – RMS™, and ESRI ArcView™ version 3.3 and 8. All purchased plug-ins are preloaded on the ViewPoint Appliance prior to delivery. Additional Input Device plug-ins, Client plug-ins, and User/Workstation License Bundles can be purchased and added to the original ViewPoint appliance via a patch file and updated security key
- Real-time personnel dosimetry and data management
- Area monitoring
- Integrated remote monitoring
- Instrument management
- First Responder environmental monitoring
- Site Process Monitoring
Specifications
Plume Tracker Simulator Viewpoint Plug-In | |
Windows™ NT/2000/XP | |
Real time personnel dosimetry and data management, area monitoring, integrated remote monitoring, instrument management, first responder environmental monitoring, site process monitoring |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.