Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Thermo-Start™ Taq DNA Polymerase PCR Kts and Master Mxes

Hot-start modified for PCR applications requiring maximum specificity and sensitivity to detect rare or complex targets and allow for room-temperature reaction setup.

Specifications

Compatibility Thermo-Start Taq Reddymix Kits and Master Mixes
For Use With (Application) Amplifying fragments up to 5kb; Hot-Start PCR; RT-PCR; Sequencing; Multiplexing; Genotyping; Library Construction; Cloning; Pathogen Detection; Single Cell PCR
Storage Requirements Store at -20°C for long-term storage. Stable at 4°C for up to four weeks. Avoid freeze-thaw cycles.
View More Specs

Products 1
Catalog Number Mfr. No. Description Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Description Quantity Price Quantity & Availability  
AB-0908B
View Documents
Thermo Scientific™
AB0908B
Thermo-Start DNA Polymerase Enzyme Kit 4000 x 25µL Rxns This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Thermo Scientific™ Thermo-Start mixes are hot-start modified for PCR applications requiring maximum specificity and sensitivity, to detect rare or complex targets; also allows for room-temperature reaction setup.

Thermo-Start Taq DNA Polymerase is a hot-start enzyme combined with an optimized buffer system that gives superior specificity and sensitivity. The chemically modified Thermo-Start Taq remains completely inactive until it is switched on by a high temperature incubation step at the start of the PCR reaction. This prevents nonspecific amplification during reaction setup, eliminating smearing and the formation of nontarget bands.
  • Hot-Start modification ensures stringent specificity
  • Safe for room temperature reaction setup
  • Enhanced enzyme stability for increased yield and sensitivity
  • Advanced buffering system minimizes PCR optimization steps
  • High Performance Buffer formulation for GC-rich and other complex templates
Thermo-Start is available in a range of formats and pack sizes to suit any workflow.

Format Options:

  • Enzyme Kits are the most basic format. Kits include the enzyme, PCR buffer, and 25mM MgCl2 solution, each in separate vials. dNTPs must be purchased separately

  • Complete PCR Kits include all the components needed to perform PCR, including an HPLC-purified dNTP mix. Complete PCR kits are a convenient option that still allows for MgCl2 optimization

  • Master Mix formats consist of a single master buffer containing all the components needed for PCR. Master Mixes are the most convenient format as they significantly reduce assay set-up steps and minimize the risk of sample contamination

  • Premixed MgCl2 Kits have MgCl2 premixed into the PCR buffer and will be at 1.5mM concentration in the final reaction

  • High Performance Buffer Kits are specially formulated for GC-rich or other complex targets with a high degree of secondary structure

Compatible with:

Thermo-Start Taq Reddymix Kits and Master Mixes

Recommended for:

Amplifying fragments up to 5kb; Hot-Start PCR; RT-PCR; Sequencing; Multiplexing; Genotyping; Library Construction; Cloning; Pathogen Detection; Single Cell PCR

Specifications

Specifications

Thermo-Start Taq Reddymix Kits and Master Mixes
Amplifying fragments up to 5kb; Hot-Start PCR; RT-PCR; Sequencing; Multiplexing; Genotyping; Library Construction; Cloning; Pathogen Detection; Single Cell PCR
Store at -20°C for long-term storage. Stable at 4°C for up to four weeks. Avoid freeze-thaw cycles.
SDS

Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser's own internal research. No other patent rights are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit