Learn More
Thermo Scientific™ ThermoChill I Recirculating Chillers
Compact line of refrigerated recirculators ranging in cooling capacities from 600 to 2000 watts
Manufacturer:  Thermo Scientific™ 196111010000
 View more versions of this product
Catalog No. 13-875-100
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Up to 700 Watts
±0.1°C
R134A
CE marked /UL listed
PD 1 - Positive Displacement
115V/60Hz
ThermoChill I, 115V/60Hz, PD-1
5 to +30°C
1.4 gpm @ 60 psid (5.3 lpm @ 4.1 bar)
None