Thermo Scientific™ ThermoChill II Low Range Recirculating Chillers
Compact line of refrigerated recirculators offering cooling capacities up to 2000 watts
Manufacturer: Thermo Scientific™ 197213010000
 View more versions of this product
Catalog No. 13-875-107
  Check Availability
Add to Cart
    Specifications
    Flow Rate 2.0 gpm @ 6 psid (7.6 lpm @ 0.4 bar)
    Pump Type Centrifugal Pump Magnetically Driven (MD)
  • Description & Specifications
Related Products
Product Content Feedback