Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Thermogreen™ LB-1 Septa, Solid Disc

Suitable for conveniently introducing the sample without causing the system to leak. Improper septa decompose under the high-temperature conditions of capillary inlets causing baseline disruption and ghost peaks in a chromatogram.

$183.00 - $190.00

Specifications

Color Green
For Use With (Application) Gas Chromatography
No. per Pack 50/Pk.
Style Solid Disc
Temperature Range (Metric) 50°C to 300°C
View More Specs

Products 2
Catalog Number Mfr. No. Diameter (Metric) Outer Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Outer Price Quantity & Availability  
11-101-0520
View Documents
MilliporeSigma™ Supelco™
20658
11 mm
Each for $183.00
Add to Cart
 
11-101-0539
View Documents
MilliporeSigma™ Supelco™
20661
12.5 mm
Each for $190.00
Add to Cart
 
Description

Description

For use with:

  • Minimizing extraction onto or loss of analytes off the solid phase microextraction (SPME) fiber from the vial during analysis by GC-MS
  • Avoiding carrier gas leaks, extraneous peaks, and phthalate contamination in GC instruments
  • Use in conjunction with a thick needle protector to avoid damage to GC instruments irreversibly caused by microsyringes
  • Influent sampling ports in gas chromatographs
Specifications

Specifications

Green
50/Pk.
50°C to 300°C
Septa, LB-1
Gas Chromatography
Solid Disc
Thermogreen
Videos
SDS
Documents

Documents

Product Certifications

Thermogreen is a registered trademark of Sigma-Aldrich Co. LLC

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.