Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ThermoMixer™ C

Click to view available options
Temperature Range (Metric):
15°C to 100°C

Improves assay results by dry incubation and mixing of samples simultaneously. Eppendorf™ ThermoMixer™ C has anti-spill technology which prevents lid wetting and cross-contamination.

  • Mixing, heating, and cooling for high flexibility
  • SmartBlocks for all common lab vessels and plate formats from 5μL to 50mL used for standard applications
  • 2DMix-Control technology results in sample mixing
  • Temperature management for maximum temperature accuracy, resulting in safe handling
  • Compatible with Eppendorf ThermoTop (condens.protect™) for condensation-free sample handling
  • Predefined program keys and clearly arranged menu guides enable simple and intuitive operations
  • Freely programmable program keys for individual optimization
  • Eppendorf PhysioCare Concept™ provides ergonomic design and operation

Applications:

  • Plasmid/RNA/DNA purification
  • cDNA synthesis
  • Extraction of DNA fragments from agarose gels
  • Enzymatic reactions (ex: DNA restriction digestion, Proteinase K digestion and ligation)
  • Transformation of bacteria strains and plasmids
  • Denaturation of DNA, RNA, and proteins
  • Labeling of DNA, RNA, and proteins
  • Bacterial growth in microreaction vessels, conical tubes, and deepwell plates
  • Lysis reactions at 100°C
  • Mixing of PCR setups
Type ThermoMixer
Depth (Metric) 30.4 cm
Temperature Range (Metric) 15°C to 100°C
Frequency 50/60 Hz
Height (Metric) 13.6 cm
Includes 24 x 1.5mL SmartBlocks
Shipping Weight (Metric) 6.3 kg
Speed 3000rpm
Timer 15 sec. to 99:30 hour, continuous
Voltage 120 V
Wattage 200 w
Width (Metric) Platform 20.6 cm
Show More Show Less