Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ ThermoMixer™ C PROMO

Perfect choice for mixing and heating established routine applications

Manufacturer:  Eppendorf™ 2231000667

Catalog No. 05-412-503PM

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • Heating and mixing in 0.5/1.5/2.0 mL reaction vessels (F0.5/F1.5/F2.0) or plates (FP)
  • Efficient mixing up to 1,500 rpm (F1.5/F2.0) or 2,000 rpm (F0.5/FP)
  • Excellent mixing performance due to unique 2DMix-Control technology
  • Anti-spill technology prevents lid wetting and cross-contamination
  • Reliable prevention of condensate formation with the new ThermoTop (condens.protect™)
  • Simple and intuitive operation using predefined temperature keys (37 °C, 42 °C, 56 °C, and 95 °C)
  • Very quiet operation via precise fit of vessels in block
  • Heat-insulated Eppendorf SmartBlocks for ergonomic operation - user won't burn fingers
Specifications

Specifications

Max. ±0.5 °C at 20 – 45 °C
1°C to 100°C
Eppendorf SmartBlocks (sold separately)
6.3 kg (without accessories)
3000rpm
200 w
30.4 x 20.6 x 13.6 cm (11.96 x 8.11 x 5.35 in.)
100/130V, 50/60Hz
ThermoMixer C
13.88 lb. (without accessories)
15 sec to 99:30 h, continuous
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit