Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ThermoPrime™ TaqDNA Polymerase PCR Kts and Master Mixes

For PCR applications requiring increased yield and sensitivity, these PCR kits and master mixes are optimized to produce large amounts of PCR product.

Specifications

For Use With (Application) Amplifying fragments up to 5kb; PCR; RT-PCR; Sequencing; Genotyping; Library Construction; Cloning; Pathogen Detection
Storage Requirements Store at -20°C for long-term storage. Stable at 4°C for up to four weeks. Avoid freeze-thaw cycles.
View More Specs

Products 1
Catalog Number Mfr. No. Includes Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Includes Quantity Price Quantity & Availability  
AB1301B
View Documents
Thermo Scientific™
AB1301B
Enzyme Kit, Pre-Mixed MgCl2 4000 x 25µL Rxns This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Part Not Found
N/A
Description

Description

Thermo Scientific™ ThermoPrime mixes are optimized for PCR applications requiring increased yield and sensitivity, to produce large amounts of PCR product.

ThermoPrime Taq DNA Polymerase is a 94 kDa recombinant thermostable DNA polymerase obtained by high level expression of the Taq DNA polymerase gene in E. coli. A proprietary buffer component protects ThermoPrime during high temperature steps, ensuring optimal performance throughout the reaction, leading to increased PCR yields. Convenient Mater Mix format is also available for easier reaction set-up.

Highlights:
  • Enhanced enzyme stability for increased yield
  • Advanced buffering system minimizes PCR optimization steps
  • Convenient Master Mix formats save time and reduce the risk of sample contamination during assay setup

Recommended for:

Amplifying fragments up to 5kb; Routine PCR

Specifications

Specifications

Amplifying fragments up to 5kb; PCR; RT-PCR; Sequencing; Genotyping; Library Construction; Cloning; Pathogen Detection
Store at -20°C for long-term storage. Stable at 4°C for up to four weeks. Avoid freeze-thaw cycles.

Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser's own internal research. No other patent rights are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit