Learn More
Sonoco™ ThermoSafe Diagnostic Shipper, Three Tube Mailer System Available on GSA/VA Contract for Federal Government customers only.
Mailer secures up to three 10mL tubes
Manufacturer:  Sonoco™ 473
 View more versions of this product
Catalog No. 03-527D
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Expanded Polystyrene (EPS) Foam
10mL Vials
Three-tube mailer system with tubes
Meets U.S. Postal Regulations, DOT regulations, and IATA Packaging Instruction 650; tubes and shipping system validated for the transport of diagnostic samples
3 x 10mL Tubes
Accessories