Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sonoco™ ThermoSafe ™ Insulated EPS Shippers Available on GSA/VA Contract for Federal Government customers only.

EPS foam corrugate shippers for the safe storage and delivery of most refrigerated or frozen products

Manufacturer:  Sonoco™ 635UPS

 View more versions of this product

Catalog No. 03-001-235


Add to cart

Description

Description

  • EPS foam containers in 200 lb. test corrugated carton
  • Excellent dry ice sublimation: average dry ice usage in 1.5 in. thick wall units is 5 pounds over 24 hours and 3-4 pounds thereafter
  • Standard wall shippers 1.5 in. thick - 24 to 48 hours time range
  • Thin wall shippers 0.75 to 1.125 in.thick  - 24 hours or less time range
  • Thick wall shippers 1.75 to 2.25 in.thick - 48 hours or more time range

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Clinical trials, diagnostic, distribution, lab supplies, medical transport short-term
I.D.: 16.125 x 11.5 x 6 in.
24 to 48 hr.
1.5 in.
Corrugate
O.D.: 19.25 x 14.5 x 9 in.
EPS Foam
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit