Learn More
Sonoco™ Insulated Shipper Multipurpose Containers and Accessories Available on GSA/VA Contract for Federal Government customers only.
Ideal shippers for the safe storage and transport of most refrigerated or frozen materials
Manufacturer:  Sonoco™ 433
 View more versions of this product
Catalog No. 03-525-43
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Expanded Polystyrene (EPS) foam assembled in corrugated carton
27.9cm
51.4cm
35.2cm
51.4 x 35.2 x 27.9cm
11 in.
20.25 in.
13.87 in.
Use for culture packs, serums, vaccines, and laboratory specimens.
Foam shipper
Accessories