Learn More
Sonoco™ Insulated Shipper Multipurpose Containers and Accessories Available on GSA/VA Contract for Federal Government customers only.
Ideal shippers for the safe storage and transport of most refrigerated or frozen materials
Manufacturer:  Sonoco™ 479
 View more versions of this product
Catalog No. 03-528-27
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Expanded Polystyrene (EPS) foam; Dome style mailer assembled in corrugated carton
20cm
39cm
29cm
39 x 29 x 20cm
8 in.
15.5 in.
11.5 in.
Use for culture packs, serums, vaccines, and laboratory specimens.
Insulated Shipper
Accessories