Learn More
Sonoco™ ThermoSafe LabMailer Specimen Mailer
Lightweight, economical lab mailer shippers
Supplier:  Sonoco™ 602
 View more versions of this product
Catalog No. 03-525-6
Add to Cart
Description
Description
  • Hold screw-capped or rubber-capped tubes or vials and swabs
  • Provide maximum breakage protection for known nonhazardous materials
  • Protect frozen, refrigerated, and ambient specimens required for compliance, analysis, testing, research and development, public health, industrial hygiene and environmental use
Additional packaging may be required to conform to CFR72.2 specification (See Packaging Accessories Kit Mfr. No. 471).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Corrugated carton for Model 600
Model 600
LabMailer
Product Suggestions
Product Suggestions
SDS
Documents
Documents
Product Certifications
Accessories