Learn More
Sonoco™ ThermoSafe LabMailer Specimen Tube Mailers
Shock-absorbing polystyrene foam mailers accept sample tubes with rubber or screw caps
Supplier: Sonoco™ 362
Description
- Two-piece assembly with snap-lock fasteners holds contents in place
- Ideal for mailing or transporting nonhazardous blood, urine, or other lab samples.
Specifications
2 Tubes | |
5.5 x 3.5 x 1.75 in. | |
107 mm | |
5, 7, or 10 mL |
Ideal for mailing or transporting nonhazardous blood, urine, or other lab samples | |
16 mm | |
Foam only shipper. Must purchase mailing sleeve model 363, which Holds 2 Tubes or 2 slides | |
Polystyrene Foam |