Learn More
Sonoco™ ThermoSafe Laminated Fiberboard Storage/Transport Units
Lightweight, low-cost mobile storage units. For transport of a wide variety of laboratory, field collection, and field applications. Ideal for dry ice or wet ice storage.
Manufacturer:  Sonoco™ 399
 View more versions of this product
Catalog No. 03-525-33
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Lightwieght laminated fiberboard wtih EPS foam interior.
  • Low-temperature storage chests include 124.7kg (275 lb.) bursting test laminated fiberboard exterior and expanded polystyrene foam interior; with insulated lid.
  • Units are low maintenance and reusable.
  • Available in different sizes for different holding time.
  • Straps, harness, and dollies are sold separately.
Order Info
Dolly not included with chests
Specifications
Specifications
1.5 cu. ft.
Hard-Cased Chest (no handles)
Included with carrying harness, no handle
6.5 days
81 lb.
SDS
Documents
Documents
Product Certifications
Accessories