Learn More
Sonoco™ ThermoSafe™ PolarPack™ Foam Bricks
Vacuum-sealed for consistent weight and shape. ThermoSafe™ PolarPack™ semi-rigid foam bricks never change shape during freezing, thawing or transit, enabling superior surface contact for more consistent temperature control.
$31.50 - $93.10
Specifications
Applications Non-mechanical refrigeration
Description Refrigerant Pack
Material Semi-Rigid Foam
Type PolarPack Foam Brick
View More Specs
Products 11
Catalog Number Mfr. No. Weight (English) Dimensions (L x W x H) Price Quantity & Availability  
Catalog Number Mfr. No. Weight (English) Dimensions (L x W x H) Price Quantity & Availability  
03-600-178 View Documents Sonoco™
FPP39
39 oz. 9 x 9 x 0.75 in. / 22.9 x 22.9 x 1.9 cm Pack of 14 for $81.00
Add to cart
03-600-250 View Documents Sonoco™
FPP5
5 oz. 4.5 x 4 x 0.5 in. / 11.4 x 10.1 x 1.3 cm Pack of 102 for $93.10
Add to cart
03-600-251 View Documents Sonoco™
FPP9
9 oz. 4.5 x 4 x 1 in. / 11.4 x 10.1 x 2.5 cm Pack of 48 for $53.30
Add to cart
03-600-252 View Documents Sonoco™
FPP17
17 oz. 9 x 4.75 x 0.75 in. / 22.8 x 12.1 x 1.9 cm Pack of 32 for $45.00
Add to cart
03-600-253 View Documents Sonoco™
FPP20
20 oz. 7 x 5 x 1 in. / 17.8 x 12.7 x 2.5 cm Pack of 24 for $37.75
Add to cart
03-600-254 View Documents Sonoco™
FPP22
22 oz. 6 x 6 x 1.25 in. / 15.2 x 15.2 x 3.175 cm Pack of 20 for $37.05
Add to cart
03-600-255 View Documents Sonoco™
FPP26
26 oz. 10.75 x 9.25 x 0.5 in. / 27.3 x 23.5 x 1.3 cm Pack of 24 for $45.75
Add to cart
03-600-256 View Documents Sonoco™
FPP27
27 oz. 7.75 x 7.75 x 0.875 in. / 19.7 x 19.7 x 2.2 cm Pack of 18 for $46.30
Add to cart
03-600-259 View Documents Sonoco™
FPP61
61 oz. 10.5 x 7.5 x 1.375 in. / 26.7 x 19.1 x 3.5 cm Pack of 8 for $31.50
Add to cart
03-600-258 View Documents Sonoco™
FPP38
38 oz. 10.75 x 9.25 x 0.75 in. / 27.3 x 23.5 x 1.9 cm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
03-600-257 View Documents Sonoco™
FPP30
30 oz. 5.5 x 8.5 x 1.125 in. / 14 x 21.6 x 2.8 cm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
PolarPack™ refrigerants provide thermal solutions for a vast array of temperature critical packages during transit.
  • Available in a broad selection of sizes
  • Reliable non-mechanical temperature protection
  • Foam bricks never change shape to maintain packing configuration throughout the trip
Specifications
Specifications
Non-mechanical refrigeration
Refrigerant Pack
Semi-Rigid Foam
PolarPack Foam Brick