Learn More
Sonoco™ ThermoSafe™ PureTemp™ Phase Change Materials Available on GSA/VA Contract for Federal Government customers only.
PureTemp™ Phase Change Materials (PCM) liquefy or solidify at precise temperatures. Pre-certified for frozen (in foam bricks) and control room temperature (in hard bottles) shipments.
$483.50 - $1157.00
Specifications
Type Phase Change Material
Applications Maintaining temperature during shipping
View More Specs
Products 4
Catalog Number Mfr. No. Description Weight (English) For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. Description Weight (English) For Use With (Equipment) Price Quantity & Availability  
03-600-180 Available on GSA/VA Contract for Federal Government customers only. View Documents Sonoco™
B27V
Rigid Bottle 29.3 oz. Frozen materials Pack of 16 for $559.50
Add to cart
03-600-181 View Documents Sonoco™
B38V
Rigid Bottle 45.2 oz. Frozen materials Pack of 12 for $483.50
Add to cart
03-600-191 Available on GSA/VA Contract for Federal Government customers only. View Documents Sonoco™
TPB3020Y
Foam Brick 56.0 oz. Controlled Room Temperature materials Pack of 12 for $1,157.00
Add to cart
03-600-192 Available on GSA/VA Contract for Federal Government customers only. View Documents Sonoco™
TPB3010Y
Foam Brick 38.0 oz. Controlled Room Temperature materials Pack of 16 for $927.00
Add to cart
Description
Description
When the surrounding temperature falls below the phase temperature, they solidify and release their stored energy. If the temperature rises above the phase temperature, they liquefy and absorb excess energy. These two phenomena protect the product from excessive heat and cold to help maintain product temperature for extended periods of time. PureTemp is engineered to react to a precise temperature and to operate in a very narrow temperature range.
  • Pre-qualified for refrigerated, frozen and CRT temperatures
  • Biodegradable, with USDA BioPreferred™ designation
  • Reduces shipping weight, freight, warehouse space, and packout space
  • Eliminates thermal shock with fewer excursions
  • Decreases box size and increases payload efficiency
  • Reduces shipping weight and freight costs
  • Simplifies packout inventory and process to eliminate bench-time
  • Increases shipping duration
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Phase Change Material
Maintaining temperature during shipping