Learn More
Sonoco™ ThermoSafe™ Via™ Blood Transporter
Transports sensitive bio substance safely while maintaining required temperatures. Sonoco™ ThermoSafe™ Via™ Reusable Container for Blood Transport and Storage is ideal for the transport and temporary storage of packed red blood cells, platelets, specimens, and organs.
Supplier: Sonoco™ B1R8HU
Description
Pre-qualified to maintain packed red blood cells at 1-6°C for 8 hours and 1-10°C for a minimum of 12 hours. It is ideal for the transport and temporary storage of packed red blood cells, platelets, specimens and organs.- Durable construction: Blood Transporter is reusable and will withstand years of use
- Zero Bench-Time™: refrigerant technology reduces assembly process for time and temperature critical materials
Specifications
Blood, Organ, Tissue, Specimen, Diagnostics, Lab Distribution | |
Insulated Shipper | |
16.32 lb. | |
7.4 kg | |
3-7 blood bags (250 mL each) |
Red | |
17.69 x 14.5 x 14.44 in. 44.9 x 36.8 x 36.7 cm |
|
Storage, 1 to 6°C: 8 hours; Transport, 1 to 10°C: min. 12 hr. | |
9.11 x 7.3 x 6.65 in. 23.1 x 18.5 x 16.9 cm |
|
Expanded Polypropylene |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.