Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiamine Hydrochloride, U.S.P. - F.C.C., J.T.Baker™

Click to view available options
Quantity:
100g
2.5kg
Packaging:
Poly Pail
Wide Mouth Amber Glass Bottle
67-03-8
C12H18Cl2N4OS
337.263
DPJRMOMPQZCRJU-UHFFFAOYSA-M
thiamine hydrochloride,vitamin b1 hydrochloride,thiamine hcl,aneurine hydrochloride,berin,thiamine chloride hydrochloride,trophite,thiamin chloride,thiamine dichloride,thiaminium chloride
6202
CHEBI:49105
2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;chloride;hydrochloride
CC1=C(SC=[N+]1CC2=CN=C(N=C2N)C)CCO.Cl.[Cl-]
67-03-8
C12H18Cl2N4OS
337.263
DPJRMOMPQZCRJU-UHFFFAOYSA-M
thiamine hydrochloride,vitamin b1 hydrochloride,thiamine hcl,aneurine hydrochloride,berin,thiamine chloride hydrochloride,trophite,thiamin chloride,thiamine dichloride,thiaminium chloride
6202
CHEBI:49105
2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;chloride;hydrochloride
CC1=C(SC=[N+]1CC2=CN=C(N=C2N)C)CCO.Cl.[Cl-]
Quantity 100g
Packaging Wide Mouth Amber Glass Bottle
Formula Weight 337.27
Grade FCC/USP
Chemical Name or Material Thiamine Hydrochloride