Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thianaphthene, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
95-15-8
C8H6S
134.2
MFCD00005864
FCEHBMOGCRZNNI-UHFFFAOYSA-N
benzo b thiophene,thianaphthene,benzothiophene,benzothiofuran,thionaphthene,1-thiaindene,thianaphthen,benzothiophen,thianaphtene,1-benzothiophen
7221
CHEBI:35858
1-benzothiophene
C1=CC=C2C(=C1)C=CS2
95-15-8
C8H6S
134.2
MFCD00005864
FCEHBMOGCRZNNI-UHFFFAOYSA-N
benzo b thiophene,thianaphthene,benzothiophene,benzothiofuran,thionaphthene,1-thiaindene,thianaphthen,benzothiophen,thianaphtene,1-benzothiophen
7221
CHEBI:35858
1-benzothiophene
C1=CC=C2C(=C1)C=CS2
Specific Gravity 1.14
Boiling Point 221.0°C to 222.0°C
Packaging Glass bottle
Color Pink to White or Yellow
Flash Point 110°C
Infrared Spectrum Authentic
Quantity 5g
Melting Point 28.0°C to 32.0°C
Density 1.1400g/mL
Assay Percent Range 97%
Beilstein 17, 59
Merck Index 15, 9455
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in usual organic solvents, soluble in ethanol, ethylether, acetone
Formula Weight 134.2
Physical Form Crystalline Solid
Percent Purity 97%
Chemical Name or Material Thianaphthene
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning