Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiazolidine, 97%, Thermo Scientific Chemicals

Catalog Number AAH2642703
Click to view available options
Quantity:
1 g
5 g
504-78-9
C3H8NS
90.16
MFCD00005211
OGYGFUAIIOPWQD-UHFFFAOYSA-O
thiazolidine, tetrahydrothiazole, 1-thia-3-azacyclopentane, thiazole, tetrahydro, unii-i320806bkw, ccris 4275, acmc-1ao0m, 4-27-00-00009 beilstein handbook reference
10444
CHEBI:50120
1,3-thiazolidine
C1CSC[NH2+]1
504-78-9
C3H8NS
90.16
MFCD00005211
OGYGFUAIIOPWQD-UHFFFAOYSA-O
thiazolidine, tetrahydrothiazole, 1-thia-3-azacyclopentane, thiazole, tetrahydro, unii-i320806bkw, ccris 4275, acmc-1ao0m, 4-27-00-00009 beilstein handbook reference
10444
CHEBI:50120
1,3-thiazolidine
C1CSC[NH2+]1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.131
Boiling Point 165°C
Flash Point 56°C (133°F)
Odor Characteristic
Refractive Index 1.5545
Quantity 1 g
UN Number UN1993
Formula Weight 89.16
Percent Purity 97%
Chemical Name or Material Thiazolidine
Hazard Category H226-H314
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 208-002-5
RTECSNumber XJ5123700
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit