Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thimerosal, 97.0 to 101.0% (Dry Basis), Affymetrix/USB™

Click to view available options
Quantity:
100g
Packaging:
Plastic bottle
54-64-8
C9H9HgNaO2S
404.811
MFCD00013062
RTKIYNMVFMVABJ-UHFFFAOYSA-L
thimerosal,thiomersal,thiomersalate,mercurothiolate,sodium merthiolate,sodium ethylmercurithiosalicylate,merthiolate,thimerosalate,thimerosalum,thimersalate
16684434
CHEBI:9546
sodium;(2-carboxylatophenyl)sulfanyl-ethylmercury
CC[Hg]SC1=CC=CC=C1C(=O)[O-].[Na+]
54-64-8
C9H9HgNaO2S
404.811
MFCD00013062
RTKIYNMVFMVABJ-UHFFFAOYSA-L
thimerosal,thiomersal,thiomersalate,mercurothiolate,sodium merthiolate,sodium ethylmercurithiosalicylate,merthiolate,thimerosalate,thimerosalum,thimersalate
16684434
CHEBI:9546
sodium;(2-carboxylatophenyl)sulfanyl-ethylmercury
CC[Hg]SC1=CC=CC=C1C(=O)[O-].[Na+]
Quantity 100g
Melting Point 234°C to 237°C
Packaging Plastic bottle
Flash Point ≥110°C (230°F)
UN Number UN2025
Formula Weight 404.81
Physical Form Powder
Percent Purity 97.0 to 101.0%
Chemical Name or Material Thimerosal
Assay Dry basis