Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thimerosal, USP, EP, BP, 97-102%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle
54-64-8
C9H9HgNaO2S
404.81
RTKIYNMVFMVABJ-UHFFFAOYSA-L
sodium [(2-carboxylatophenyl)sulfanyl](ethyl)mercury
[Na+].CC[Hg]SC1=CC=CC=C1C([O-])=O
54-64-8
C9H9HgNaO2S
404.81
RTKIYNMVFMVABJ-UHFFFAOYSA-L
sodium [(2-carboxylatophenyl)sulfanyl](ethyl)mercury
[Na+].CC[Hg]SC1=CC=CC=C1C([O-])=O

TH125, 54-64-8, C9H9HgNaO2S

Spectrum™ Thimerosal, USP, EP, BP is used as a preservative in vaccines. All Spectrum Chemical USP, EP, BP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

CAS Max % 1
Loss on Drying 0.005
Packaging Amber Glass Bottle
Percent Purity 97 to 102%
Grade Multi Compendia
pH 6.0 to 8.0
Quantity 100 g