Learn More
Greiner Bio-One™ ThinCert™ CellCoat™ 24 Well Cell Culture Inserts
Suitable for transport, secretion, diffusion study
Manufacturer:  Greiner Bio-One™ 66265406
 View more versions of this product
Catalog No. 07-001-012
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • PET porous membrane
  • Increases cell growth and differentiation
  • Coated with collagen type 1
  • Applications:
  • Cell migration, transepithelial transport and paracrine cell-cell interactions co-cultures, ti
Specifications
Specifications
Clear
Yes
6 pcs/ 24 well plate
Collagen Type I protein
ThinCert™: 0.1 to 0.35mL; well: 0.4 to 1.2mL
33.6 sq. mm.
PET, PET
0.4μm
− 20° to 60°C
Sterile