Learn More
Thioacetamide, ACS, 99% min, Thermo Scientific Chemicals
Description
Thioacetamide is used in qualitative inorganic analysis as an in-situ source for sulfide ions. It is used in analytical chemistry as a source of hydrogen sulfide. It is an additive in enantioselective reduction of beta-keto esters with immobilized bakers yeast. It is also used as a vulcanizing agent of polymer and a crosslinking agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 112°C to 114°C |
Quantity | 10 g |
Beilstein | 506006 |
Merck Index | 14,9319 |
Solubility Information | Soluble in ethanol,benzene and petroleum ether. Slightly soluble in water and ether. |
Formula Weight | 75.13 |
Percent Purity | 99% |
Grade | ACS |
Physical Form | Crystalline |
Chemical Name or Material | Thioacetamide |
Safety and Handling
Hazard Category | H302-H315-H319-H336-H351-H373 |
Hazard Statement | GHS H Statement H350-H302-H315-H319 May cause cancer. Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. |
Precautionary Statement | P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c |
EINECSNumber | 200-541-4 |
RTECSNumber | AC8925000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.