Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thioacetamide, Crystal, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
500g
Packaging:
Wide Mouth Amber Glass Bottle
62-55-5
C2H5NS
75.129
YUKQRDCYNOVPGJ-UHFFFAOYSA-N
thioacetamide,acetothioamide,thiacetamide,acetamide, thio,thioacetimidic acid,acetimidic acid, thio,usaf cb-21,usaf ek-1719,rcra waste number u218,acetic acid, thiono-, amide
2723949
CHEBI:32497
ethanethioamide
CC(=S)N
62-55-5
C2H5NS
75.129
YUKQRDCYNOVPGJ-UHFFFAOYSA-N
thioacetamide,acetothioamide,thiacetamide,acetamide, thio,thioacetimidic acid,acetimidic acid, thio,usaf cb-21,usaf ek-1719,rcra waste number u218,acetic acid, thiono-, amide
2723949
CHEBI:32497
ethanethioamide
CC(=S)N

High quality chemicals for laboratory and specialized industrial use

Packaging Wide Mouth Amber Glass Bottle
Quantity 125g
Formula Weight 75.13
Physical Form Crystals
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Thioacetamide