Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thioacetamide (Crystalline Powder/Certified ACS), Fisher Chemical

Click to view available options
Quantity:
100g
500g
Packaging:
Glass Bottle
62-55-5
C2H5NS
75.129
MFCD00008070
YUKQRDCYNOVPGJ-UHFFFAOYSA-N
thioacetamide,acetothioamide,thiacetamide,acetamide, thio,thioacetimidic acid,acetimidic acid, thio,usaf cb-21,usaf ek-1719,rcra waste number u218,acetic acid, thiono-, amide
2723949
CHEBI:32497
ethanethioamide
CC(=S)N
62-55-5
C2H5NS
75.129
MFCD00008070
YUKQRDCYNOVPGJ-UHFFFAOYSA-N
thioacetamide,acetothioamide,thiacetamide,acetamide, thio,thioacetimidic acid,acetimidic acid, thio,usaf cb-21,usaf ek-1719,rcra waste number u218,acetic acid, thiono-, amide
2723949
CHEBI:32497
ethanethioamide
CC(=S)N
Quantity 500g
pH 5.2
Identification Pass Test
Packaging Glass Bottle
Ignition Residue ≤0.05%
Melting Point 112°C
Color Cream
Appearance Pass Test (2% solution)
Assay Percent Range ≥99 %
Linear Formula CH3CSNH2
Formula Weight 75.13
Physical Form Solid
Percent Purity ≥99%
Grade Certified ACS
Chemical Name or Material Thioacetamide
Show More Show Less