Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thioanisole 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
2.5kg
Packaging:
Glass bottle
100-68-5
C7H8S
124.21
MFCD00008559
HNKJADCVZUBCPG-UHFFFAOYSA-N
thioanisole,methyl phenyl sulfide,methylthio benzene,thioanisol,benzene, methylthio,phenylthiomethane,phenyl methyl sulfide,methylsulfanyl benzene,methylphenylsulfide,1-thiaethyl benzene
7520
methylsulfanylbenzene
CSC1=CC=CC=C1
100-68-5
C7H8S
124.21
MFCD00008559
HNKJADCVZUBCPG-UHFFFAOYSA-N
thioanisole,methyl phenyl sulfide,methylthio benzene,thioanisol,benzene, methylthio,phenylthiomethane,phenyl methyl sulfide,methylsulfanyl benzene,methylphenylsulfide,1-thiaethyl benzene
7520
methylsulfanylbenzene
CSC1=CC=CC=C1
Specific Gravity 1.05
Water 0.5% max.
Density 1.0500g/mL
Quantity 500g
Boiling Point 188.0°C
Flash Point 73°C
Color Brown-Yellow to Yellow
Packaging Glass bottle
Melting Point -15.0°C
Refractive Index 1.5842 to 1.5862
Infrared Spectrum Authentic
Assay Percent Range 99%
Linear Formula C6H5SCH3
Beilstein 06, 297
Fieser 05,660; 06,582
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 124.21
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Thioanisole, 99%
Show More Show Less

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-878-2

RTECSNumber : DA6200000

TSCA : TSCA