Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiobenzoic acid, 90%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
98-91-9
C7H6OS
138.18
MFCD00004852
UIJGNTRUPZPVNG-UHFFFAOYSA-N
thiobenzoic acid,benzenecarbothioic acid,benzoyl thiol,monothiobenzoic acid,benzoic acid, thio,acido mercaptobenzoico,unii-gbg5rlo56n,ccris 8913,acido mercaptobenzoico italian,gbg5rlo56n
7414
benzenecarbothioic S-acid
C1=CC=C(C=C1)C(=O)S
98-91-9
C7H6OS
138.18
MFCD00004852
UIJGNTRUPZPVNG-UHFFFAOYSA-N
thiobenzoic acid,benzenecarbothioic acid,benzoyl thiol,monothiobenzoic acid,benzoic acid, thio,acido mercaptobenzoico,unii-gbg5rlo56n,ccris 8913,acido mercaptobenzoico italian,gbg5rlo56n
7414
benzenecarbothioic S-acid
C1=CC=C(C=C1)C(=O)S
Specific Gravity 1.17
Melting Point 15.0°C to 18.0°C
Boiling Point 122.0°C (30.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Color Yellow to Brown
Density 1.1700g/mL
Packaging Glass bottle
Refractive Index 1.6020 to 1.6080
Quantity 25g
Assay Percent Range 90%
Linear Formula C6H5COSH
Beilstein 09, 419
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with organic solvents
Formula Weight 138.18
Physical Form Liquid
Percent Purity 90%
Chemical Name or Material Thiobenzoic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.