Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Thiobenzoic acid, 94%

Click to view available options
Quantity:
100g
98-91-9
C7H6OS
138.184
MFCD00004852
UIJGNTRUPZPVNG-UHFFFAOYSA-N
thiobenzoic acid,benzenecarbothioic acid,benzoyl thiol,monothiobenzoic acid,benzoic acid, thio,acido mercaptobenzoico,unii-gbg5rlo56n,ccris 8913,acido mercaptobenzoico italian,gbg5rlo56n
7414
benzenecarbothioic S-acid
C1=CC=C(C=C1)C(=O)S
98-91-9
C7H6OS
138.184
MFCD00004852
UIJGNTRUPZPVNG-UHFFFAOYSA-N
thiobenzoic acid,benzenecarbothioic acid,benzoyl thiol,monothiobenzoic acid,benzoic acid, thio,acido mercaptobenzoico,unii-gbg5rlo56n,ccris 8913,acido mercaptobenzoico italian,gbg5rlo56n
7414
benzenecarbothioic S-acid
C1=CC=C(C=C1)C(=O)S
Applications
Thiobenzoic acid is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
Quantity 100g
Odor Stench
Melting Point 15°C to 18°C
Flash Point >110°C (230°F)
Boiling Point 103°C to 105°C (12mmHg)
Beilstein 1071790
Formula Weight 138.19
Percent Purity 94%
Chemical Name or Material Thiobenzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : DH6839000

TSCA : TSCA

Recommended Storage : 2° to 8°C