Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiocarbanilide 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
1kg
2.5kg
Packaging:
Plastic bottle
102-08-9
C13H12N2S
228.32
MFCD00004921
FCSHMCFRCYZTRQ-UHFFFAOYSA-N
thiocarbanilide,diphenylthiourea,1,3-diphenyl-2-thiourea,n,n'-diphenylthiourea,sulfocarbanilide,sym-diphenylthiourea,thiourea, n,n'-diphenyl,stabilisator c,rhenocure ca,vulkacit ca
700999
1,3-diphenylthiourea
C1=CC=C(C=C1)NC(=S)NC2=CC=CC=C2
102-08-9
C13H12N2S
228.32
MFCD00004921
FCSHMCFRCYZTRQ-UHFFFAOYSA-N
thiocarbanilide,diphenylthiourea,1,3-diphenyl-2-thiourea,n,n'-diphenylthiourea,sulfocarbanilide,sym-diphenylthiourea,thiourea, n,n'-diphenyl,stabilisator c,rhenocure ca,vulkacit ca
700999
1,3-diphenylthiourea
C1=CC=C(C=C1)NC(=S)NC2=CC=CC=C2
Quantity 100g
Infrared Spectrum Authentic
Loss on Drying 0.3% max. (60°C)
Color White to Yellow
Packaging Plastic bottle
Melting Point 151.0°C to 154.0°C
Assay Percent Range 98%
Linear Formula C6H5NHCSNHC6H5
Beilstein 12,394
Merck Index 15,3373
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, ether and chloroform
Formula Weight 228.32
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material Thiocarbanilide, 98%
Show More Show Less

GHS P Statement May cause an allergic skin reaction. Suspected of damaging the unborn child. May cause damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

Warning

EINECSNumber : 203-004-2

RTECSNumber : FE1225000

TSCA : TSCA