Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiocarbohydrazide, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
Plastic bottle
2231-57-4
CH6N4S
106.15
MFCD00007616
LJTFFORYSFGNCT-UHFFFAOYSA-N
thiocarbohydrazide,thiocarbazide,thiocarbonohydrazide,carbonothioic dihydrazide,carbohydrazide, thio,thiocarbonic dihydrazide,1,3-diamino-2-thiourea,hydrazinecarbohydrazonothioic acid,usaf ek-7372,unii-1iz2h82nwu
2724189
CHEBI:9549
1,3-diaminothiourea
C(=S)(NN)NN
2231-57-4
CH6N4S
106.15
MFCD00007616
LJTFFORYSFGNCT-UHFFFAOYSA-N
thiocarbohydrazide,thiocarbazide,thiocarbonohydrazide,carbonothioic dihydrazide,carbohydrazide, thio,thiocarbonic dihydrazide,1,3-diamino-2-thiourea,hydrazinecarbohydrazonothioic acid,usaf ek-7372,unii-1iz2h82nwu
2724189
CHEBI:9549
1,3-diaminothiourea
C(=S)(NN)NN
Infrared Spectrum Authentic
Packaging Plastic bottle
Color Beige-Gray to White
Melting Point 168.0°C to 172.0°C
Quantity 25g
Assay Percent Range 97.5% min
Linear Formula H2NNHCSNHNH2
Beilstein 03, 197
Formula Weight 106.15
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material Thiocarbohydrazide, 98%
Show More Show Less

GHS H Statement
Fatal if swallowed.
Toxic in contact with skin.
Fatal if inhaled.

GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Do not eat,drink or smoke when using th

GHS Signal Word: Danger

EINECSNumber : 218-769-8

RTECSNumber : FF2975000

TSCA : TSCA