Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiodiglycolic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
1kg
Packaging:
Plastic bottle
123-93-3
C4H6O4S
150.148
MFCD00004359
UVZICZIVKIMRNE-UHFFFAOYSA-N
thiodiglycolic acid,2,2'-thiodiacetic acid,thiodiacetic acid,mercaptodiacetic acid,tdga,dicarboxymethyl sulfide,thiodi acetic acid,carboxymethylthio acetic acid,2,2'-thiodiethanoic acid,thiodiglycollic acid
31277
CHEBI:35012
2-(carboxymethylsulfanyl)acetic acid
C(C(=O)O)SCC(=O)O
123-93-3
C4H6O4S
150.148
MFCD00004359
UVZICZIVKIMRNE-UHFFFAOYSA-N
thiodiglycolic acid,2,2'-thiodiacetic acid,thiodiacetic acid,mercaptodiacetic acid,tdga,dicarboxymethyl sulfide,thiodi acetic acid,carboxymethylthio acetic acid,2,2'-thiodiethanoic acid,thiodiglycollic acid
31277
CHEBI:35012
2-(carboxymethylsulfanyl)acetic acid
C(C(=O)O)SCC(=O)O
Assay Percent Range 98%
Linear Formula S(CH2CO2H)2
Beilstein 03, 253
Merck Index 15, 9484
Formula Weight 150.15
Physical Form Crystalline Powder
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Plastic bottle
Solubility Solubility in water: soluble. Other solubilities: soluble in alcohol, insoluble in benzene
Melting Point 128.0°C to 131.0°C
Color Gray to White
Quantity 1kg
Chemical Name or Material Thiodiglycolic acid, 98%
Show More Show Less

GHS P Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 204-663-9

RTECSNumber : AJ6475000

TSCA : TSCA