Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thioflavin S, Acros Organics™

Click to view available options
Quantity:
5g
25g
100g
1326-12-1
C.I. 49010,Direct Yellow 7
1326-12-1
C.I. 49010,Direct Yellow 7
Lambda Max 360 to 380nm
Quantity 25g
Chemical Name or Material Thioflavin S

GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement IF ON SKIN: Wash with plenty of soap and water. If eye irritation persists: Get medical advice/attention. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection. If skin irritation occurs: Get medical advice/attention.

Warning